Merck
  • A case of acute prochloraz-manganese complex intoxication treated with extracorporeal elimination.
A case of acute prochloraz-manganese complex intoxication treated with extracorporeal elimination.
Blood purification (2013-01-12)
Hyowook Gil, Sujin Seok, Jungrak Hong, Mihye Jeong, Saeyong Hong
ABSTRACT
We treated a patient with critical manganese intoxication with vigorous extracorporeal elimination. In this article, we describe the clinical characteristics and treatment modalities of the patient. A 65-year-old man was brought to the emergency room (ER) 5.5 h after ingesting prochloraz-manganese complex. He experienced circulatory collapse and went into a coma without self-breathing on arrival at the ER. Mechanical ventilation was initiated and hemoperfusion, hemodialysis and continuous venovenous hemodiafiltration were performed with the help of norepinephrine. MEASUREMENT AND RESULT: The manganese levels on the first, second and fourth hospital days were 34.1, 23.6 and 12.5 µg/l, respectively. He recuperated from the shock state within 7 hospital days. After 4 critical weeks, the patient regained full consciousness. Rigorous extracorporeal elimination by hemoperfusion, hemodialysis and continuous venovenous hemodiafiltration was an effective treatment modality for patients with acute manganese intoxication.
Product Description
Supelco
Prochloraz, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Prochloraz, PESTANAL®, analytical standard