Merck
  • [Experimental justification of approaches to pharmacological correction of delayed disorders caused by acute ethylene glycol poisoning].
[Experimental justification of approaches to pharmacological correction of delayed disorders caused by acute ethylene glycol poisoning].
Eksperimental'naia i klinicheskaia farmakologiia (2013-01-18)
M M Liubishin, K V Sivak, T N Savateeva-Liubimova
ABSTRACT
The development of delayed disorders caused by acute ethylene glycol poisoning has been studied in experiments on male rats. These disorders include chronic renal failure and secondary combined immunodeficiency status of the "circulus vitiosus" type. Urgent pharmacological correction was shown to be necessary shortly after the poisoning. The experimental therapy (administration of immunomodulators with various mechanisms of action in addition to conventional antidote treatment with ethanol) resulted in the restoration of nonspecific resistance and both cellular and humoral immunity. Reduction of the urinary system damage after the administration of immunomodulators was observed. The results demonstrated the importance of multiagent immunotherapy for the correction of delayed effects of acute ethylene glycol poisoning.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
9-Oxo-10(9H)-acridineacetic acid, derivatization grade (HPLC), ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylene glycol 5 M solution