Merck
  • Immunomodulation and the quorum sensing molecule 3-oxo-C12-homoserine lactone: the importance of chemical scaffolding for probe development.
Immunomodulation and the quorum sensing molecule 3-oxo-C12-homoserine lactone: the importance of chemical scaffolding for probe development.
Chemical communications (Cambridge, England) (2013-01-19)
Amanda L Garner, Jing Yu, Anjali K Struss, Gunnar F Kaufmann, Vladimir V Kravchenko, Kim D Janda
ABSTRACT
As a guide for chemical probe design, focused analogue synthetic studies were undertaken upon the lactone ring of 3-oxo-C(12)-homoserine lactone. We have concluded that hydrolytic instability of the heterocyclic ring is pivotal for its ability to modulate immune signaling and probe preparation was aligned with these findings.
Sigma-Aldrich
α-Amino-γ-butyrolactone hydrobromide, 99%
Sigma-Aldrich
(S)-α-Amino-γ-butyrolactone hydrochloride, 97%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
L-Homoserine lactone hydrochloride
Sigma-Aldrich
L-Homoserine
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
Dihydro-3-amino-2-(3H)-furanone, AldrichCPR