Merck
  • Restricted outbreak of American tegumentary leishmaniasis with high microfocal transmission.
Restricted outbreak of American tegumentary leishmaniasis with high microfocal transmission.
The American journal of tropical medicine and hygiene (2013-01-23)
Alejandro J Krolewiecki, José F Gil, Marcelo Quipildor, Silvana P Cajal, Carlos Pravia, Marisa Juarez, Carlos Villalpando, Fabricio M Locatelli, Mariana Chanampa, Gabriela Castillo, María F Oreste, Carlos L Hoyos, Vanesa Negri, Julio R Nasser
ABSTRACT
Cutaneous leishmaniasis is endemic in Salta, the northwestern province of Argentina. We describe an outbreak involving five recreational hunters whose exposure was limited to several hours in a residual patch of primary forest. All patients presented with typical cutaneous lesions after a mean incubation period of 59 days (range 15-78), and one developed simultaneous mucosal involvement. Polymerase chain reaction analysis of lesions confirmed Leishmania (V.) braziliensis as the etiologic agent in three cases. All patients were cured with anti-Leishmania treatment. Entomologic surveys in the transmission area revealed a predominance of Lutzomyia neivai. This outbreak report confirms a microfocal transmission pattern of tegumentary leishmaniasis in the Americas and based on a well-determined exposure, allows the determination of incubation times for leishmaniasis caused by Leishmania braziliensis.
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)