Merck
  • CDDP supramolecular micelles fabricated from adamantine terminated mPEG and β-cyclodextrin based seven-armed poly (L-glutamic acid)/CDDP complexes.
CDDP supramolecular micelles fabricated from adamantine terminated mPEG and β-cyclodextrin based seven-armed poly (L-glutamic acid)/CDDP complexes.
Colloids and surfaces. B, Biointerfaces (2013-01-29)
Dawei Yong, Yu Luo, Fang Du, Jin Huang, Wei Lu, Zhaoyun Dai, Jiahui Yu, Shiyuan Liu
ABSTRACT
This research is aimed to develop a nano-sized supramolecular micelle delivery system of cis-dichlorodiammine platinum (II) (CDDP) in order to achieve the passive tumor targeting. Firstly, star-shaped poly (γ-benzyl-L-glutamate) was synthesized by the ring-opening polymerization of γ-benzyl-L-glutamate-N-carboxyanhydride initiated with per-6-amino-β-cyclodextrin. After removal of benzyl groups, β-cyclodextrin based seven-armed poly (L-glutamic acid) (β-CD-7PLGA) was obtained. β-CD-7PLGA/CDDP complexes were prepared by the complex reaction between the carboxylic groups of β-CD-7PLGA and CDDP. Further inclusion of β-CD-7PLGA/CDDP complexes with adamantine terminated mPEG (mPEG-Ad) gave CDDP supramolecular micelles (mPEG-Ad@β-CD-7PLGA/CDDP). The formation of mPEG-Ad@β-CD-7PLGA/CDDP supramolecular micelles was confirmed by fluorescence spectrophotoscopy and particle size measurements. All the micelles showed spherical shape, and their sizes increased from 100 to 135 nm with the increase of PLGA arm molecular weight. mPEG-Ad@CD-7PLGA/CDDP micelles showed sustained drug release profiles over 50h in PBS. Compared with CDDP, mPEG-Ad@β-CD-7PLGA/CDDP supramolecular micelles showed essential decreased cytotoxicity to KB cells, suggesting their great potential as the delivery carriers of CDDP.
Sigma-Aldrich
β-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
β-Cyclodextrin, ≥97%
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Sigma-Aldrich
1-Adamantylamine, 97%
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥95.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG, ≥98.0% (HPLC)
Sigma-Aldrich
β-Cyclodextrin, Wacker Chemie AG
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Sigma-Aldrich
Amantadine hydrochloride