Merck
  • Short communication: the effect of temperature on performance of milk ketone test strips.
Short communication: the effect of temperature on performance of milk ketone test strips.
Journal of dairy science (2013-01-30)
J Shire, J L Gordon, E L Karcher
ABSTRACT
Ketosis is estimated to affect 15% of early lactation dairy cows. A ketone test strip (Keto-Test; Elanco Animal Health, Greenfield, IN) allows producers a method to determine the concentration of β-hydroxybutyrate (BHBA) in milk to track individual animal and herd incidence of ketosis. The objective of this study was to determine the effect of altering the temperature of milk and the test strips at the time of the test on the reliability of the Keto-Test. A total of 118 Holstein cows, ranging from 5 to 17 DIM, were selected from a commercial Holstein dairy herd in Michigan. A milk sample was collected from the right rear quarter of each cow during the a.m. milking. Each sample was tested under 4 temperature conditions: (1) Keto-Test strips and milk at room temperature (RT; 24.0 ± 0.1°C; control; manufacturer's instructions), (2) cold strips (10.8 ± 0.9°C) and milk at RT, (3) cold strips and fresh milk, and (4) strips at RT and fresh milk. Milk was recorded as negative (0-99 μmol/L), weak positive (100-199 μmol/L), positive (200-499 μmol/L), or highly positive (≥ 500 μmol/L). Blood samples were collected immediately following milk collection and analyzed for BHBA concentration using a ketone test meter. Cows with blood BHBA concentration of ≥ 1,400 μmol/L were considered positive for subclinical ketosis. Accuracy of the Keto-Test strips under the 4 conditions was determined by the κ coefficient of agreement, using the result of condition 1 as the accepted true value. Additionally, sensitivity and specificity were calculated using the blood BHBA concentrations and results of each of the 4 conditions. Using the Keto-Test 60.2% of cows tested negative for milk BHBA, 24.6% tested weak positive, 14.4% tested positive, and 0.8% tested highly positive. The weighted κ coefficient of agreement between the control condition (1) and condition 2, 3, and 4 and 95% lower and upper confidence intervals were as follows: condition 2=0.71 (0.62, 0.80), condition 3=0.69 (0.60, 0.78), and condition 4=0.63 (0.54, 0.73). These results indicate good agreement between the outcome of condition 1 and conditions 2, 3, and 4. The sensitivities/specificities for 1, 2, 3, and 4 were as follows: 0.77/0.79, 0.74/0.75, 0.69/0.88, and 0.69/0.84, indicating that the test in all temperature conditions had a strong ability to detect the presence of BHBA in milk. In conclusion, the reliability of the Keto-Test strips was not dependent on the temperature of the milk or the test strips.
Sigma-Aldrich
3-Hydroxybutyric acid, 95%