Merck
  • Hybrid preconcentrator/focuser module for determinations of explosive marker compounds with a micro-scale gas chromatograph.
Hybrid preconcentrator/focuser module for determinations of explosive marker compounds with a micro-scale gas chromatograph.
Journal of chromatography. A (2013-01-30)
Gustavo Serrano, Thitiporn Sukaew, Edward T Zellers
ABSTRACT
This article describes the development and characterization of a partially selective preconcentrator/focuser (PCF) module for a field-portable micro-scale gas chromatograph (μGC) designed to rapidly determine trace levels of two vapor-phase markers of the explosive trinitrotoluene (TNT): 2,3-dimethyl-2,3-dinitrobutane (DMNB) and 2,4-dinitrotoluene (2,4-DNT). The PCF module has three primary components. The first is a high-volume sampler, comprising a resistively-heated 6-cm long stainless steel tube packed with tandem beds of the graphitized carbons Carbopack B (C-B, 30 mg) and Carbopack Y (C-Y, 15 mg), which traps the markers but permits more volatile interferences to pass through largely unretained. The second component is a microfocuser (μF), comprising a 4.2×9.8 mm Si chip containing a deep-reactive-ion-etched (DRIE) cavity packed with 2mg of C-B, a Pyrex cap, integrated heaters, and etched fluidic channels. The third component is a commercial polymer-membrane filter used as a pre-trap to remove particles and adsorbed low volatility interferences. Markers captured in the sampler are thermally desorbed and transferred to the μF, and then thermally desorbed/injected from the μF into a downstream separation (micro)column and detected. Scrubbed ambient air is used as carrier gas. The adsorbent capacities, baseline temperatures, sampling and desorption flow rates, and heating profiles were optimized for each PCF module component while minimizing the analysis time. An overall transfer efficiency of 86% was achieved at marker concentrations of ~0.2-2.6 ppb. In the final configuration the PCF module requires just 60s to collect a 1-L sample (3 L/min), focus (40 mL/min), and inject the markers (3 mL/min), producing half-maximum injection peak widths of ~2 and 5 s, and preconcentration factors of 4500 and 1800, for DMNB and 2,4-DNT, respectively.
Product Description
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Sigma-Aldrich
2,4-Dinitrotoluene, 97%
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Sigma-Aldrich
2,3-Dimethyl-2,3-dinitrobutane, 98%
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea