Merck
  • Clinical efficacy and safety of a multimodality skin brightener composition compared with 4% hydroquinone.
Clinical efficacy and safety of a multimodality skin brightener composition compared with 4% hydroquinone.
Journal of drugs in dermatology : JDD (2013-02-05)
Elizabeth T Makino, James H Herndon, Monya L Sigler, Vincent Gotz, John Garruto, Rahul C Mehta
ABSTRACT
There are numerous common skin disorders involving hyperpigmentation, including solar lentigines, postinflammatory hyperpigmentation, melasma, freckles, and dyschromia from photoaging. While these conditions are of an aesthetic nature, there is great interest in newer, safer, and more effective treatment modalities. Topical hydroquinone (HQ) has been the gold standard of skin lighteners for many years. However, regulatory authorities around the world are now questioning its safety. A randomized, double-blind, half-face study was conducted in females having moderate to severe facial hyperpigmentation to assess the efficacy and tolerability of 3 new skin brightener formulations containing SMA-432, a prostaglandin E2 inhibitor, compared with 4% HQ. Each subject was assigned 2 of the 4 test materials and was instructed to apply the product on the assigned side of the face twice daily for 12 weeks. Evaluation visits were conducted at baseline and at 4, 8, and 12 weeks. At each visit, subjects were evaluated by a blinded investigator for clinical efficacy and tolerability using grading scales. Standardized digital photography and Chroma Meter assessments were also taken. Self-assessment questionnaires were completed at weeks 4, 8, and 12. Sixty-eight Caucasian subjects (136 half faces) completed the study. All test materials significantly reduced overall hyperpigmentation and improved the Investigator's Global Hyperpigmentation Improvement rating at weeks 4, 8, and 12 compared with baseline. SMA-432 exhibited a dose-dependent improvement in hyperpigmentation. There were no major tolerability issues with any of the test materials. Self-assessments were generally favorable for all test materials. At the completion of the trial, subjects rated one of the tested multimodality brightener compositions as the most favorable product and 4% HQ as the least favorable. This study demonstrated that the new non-HQ-containing skin brightener formulations were as effective and equally well tolerated as the gold standard, 4% HQ, in females with facial hyperpigmentation.
Product Description
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%