Merck
  • Isoproterenol stimulates transient SNAP23-VAMP2 interaction in rat parotid glands.
Isoproterenol stimulates transient SNAP23-VAMP2 interaction in rat parotid glands.
FEBS letters (2013-02-06)
Taishin Takuma, Akiko Shitara, Toshiya Arakawa, Miki Okayama, Itaru Mizoguchi, Yoshifumi Tajima
ABSTRACT
The exocytosis of salivary proteins is mainly regulated by cAMP, although soluble N-ethylmaleimide-sensitive factor attachment protein receptors (SNAREs), which mediate cAMP-dependent exocytic membrane fusion, have remained unidentified. Here we examined the effect of isoproterenol (ISO) and cytochalasin D (CyD) on the level of SNARE complexes in rat parotid glands. When SNARE complexes were immunoprecipitated by anti-SNAP23, the coprecipitation of VAMP2 was significantly increased in response to ISO and/or CyD, although the coprecipitation of VAMP8 or syntaxin 4 was scarcely augmented. These results suggest that the SNAP23-VAMP2 interaction plays a key role in cAMP-mediated exocytosis from parotid glands.
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Supelco
DL-Dithiothreitol solution, 1 M in H2O