Merck
  • Fluorescence (fluidity/hydration) and calorimetric studies of interactions of bile acid-drug conjugates with model membranes.
Fluorescence (fluidity/hydration) and calorimetric studies of interactions of bile acid-drug conjugates with model membranes.
The journal of physical chemistry. B (2013-02-07)
Vedagopuram Sreekanth, Avinash Bajaj
ABSTRACT
We have studied the interactions of three bile acid-tamoxifen conjugates, lithocholic acid-tamoxifen (LA-Tam(1)-Am), deoxycholic acid-tamoxifen (DCA-Tam(2)-Am), and cholic acid-tamoxifen (CA-Tam(3)-Am), possessing 1-3 tamoxifen molecules having an amine headgroup with model DPPC membranes and compared with N-desmethylated tamoxifen (TamNHMe) using DPH based fluorescence anisotropy, Prodan based hydration, and differential scanning calorimetry studies. DPH based anisotropy studies showed that bile acid-tamoxifen conjugates increase membrane fluidity, which strongly depends on the number of tamoxifen molecules conjugated to bile acid and the percentage of doping of bile acid-tamoxifen conjugates in the DPPC membranes. The order of membrane fluidity of the coliposomes from bile acid-tamoxifen conjugates and DPPC lipids in gel phase was found to be CA-Tam(3)-Am > DCA-Tam(2)-Am > LA-Tam(1)-Am > TamNHMe. Incorporation of bile acid-tamoxifen conjugates showed an unusual complex behavior of membrane hydration, as evident from Prodan based hydration studies. Temperature dependent study showed incorporation of LA-Tam(1)-Am and DCA-Tam(2)-Am conjugates decreases membrane hydration with an increase in temperature up to the phase transition temperature (T(m)). Differential scanning calorimetry studies showed a decrease in phase transition temperature (T(m)) upon an increase in the percentage of doping of TamNHMe and CA-Tam(3)-Am, whereas LA-Tam(1)-Am and DCA-Tam(2)-Am do not cause a major change in the phase transition temperature (T(m)) of DPPC liposomes. These studies showed the differential behavior of bile acid-tamoxifen conjugates regulating the membrane fluidity, hydration, and phase transition of model membranes depending upon the percentage of doping and tamoxifen conjugation to bile acids.
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphocholine, ~99%
Sigma-Aldrich
Lithocholic acid, ≥95%
Lithocholic acid, European Pharmacopoeia (EP) Reference Standard
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)