Merck
  • Preparation of poly(phthalazinone-ether-sulfone) sponge-like ultrafiltration membrane.
Preparation of poly(phthalazinone-ether-sulfone) sponge-like ultrafiltration membrane.
Langmuir : the ACS journal of surfaces and colloids (2013-02-09)
Peiyong Qin, Xinjun Hong, M Nazmul Karim, Takuji Shintani, Jiding Li, Cuixian Chen
ABSTRACT
Poly(phthalazinone-ether-sulfone) (PPES) polymer is a relatively newly developed material with a bis(4-fluorodiphenyl) sulfone group. The formation of the PPES membrane by wet-phase inversion can proceed according to a slow or fast gelation method. These formation mechanisms were studied experimentally. The resulting membrane morphology was investigated using both optical and scanning electron micrography. The effects of PPES concentration and two additives, polyvinylpyrrolidone (PVP) and oxalic acid (OA), on the apparent viscosity and gelation rate of PPESK/NMP solutions and membrane performance have also been investigated. It was found that the gelation rate is important to obtain a sponge-like membrane structure, however favored by a fast gelation rate. The membrane obtained by a fast gelation rate showed a high pure water flux and rejection of bovine serum albumin (BSA), contrary to previous findings. On the basis of the experimental results, the actual membrane structure and pure water flux were related, and in agreement with the optical micrograph and gelation rate, respectively. The current results provide a fundamental insight in this novel copolymer, useful in future applications, especially in the membrane formation process.
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Povidone, meets USP testing specifications
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Kollidon® 25