Merck
  • Cloning, characterization, and sulfonamide and thiol inhibition studies of an α-carbonic anhydrase from Trypanosoma cruzi, the causative agent of Chagas disease.
Cloning, characterization, and sulfonamide and thiol inhibition studies of an α-carbonic anhydrase from Trypanosoma cruzi, the causative agent of Chagas disease.
Journal of medicinal chemistry (2013-02-09)
Peiwen Pan, Alane Beatriz Vermelho, Giseli Capaci Rodrigues, Andrea Scozzafava, Martti E E Tolvanen, Seppo Parkkila, Clemente Capasso, Claudiu T Supuran
ABSTRACT
An α-carbonic anhydrase (CA, EC 4.2.1.1) has been identified, cloned, and characterized from the unicellular protozoan Trypanosoma cruzi, the causative agent of Chagas disease. The enzyme (TcCA) has a very high catalytic activity for the CO2 hydration reaction, being similar kinetically to the human (h) isoform hCA II, although it is devoid of the His64 proton shuttle. A large number of aromatic/heterocyclic sulfonamides and some 5-mercapto-1,3,4-thiadiazoles were investigated as TcCA inhibitors. The aromatic sulfonamides were weak inhibitors (K(I) values of 192 nM to 84 μM), whereas some heterocyclic compounds inhibited the enzyme with K(I) values in the range 61.6-93.6 nM. The thiols were the most potent in vitro inhibitors (K(I) values of 21.1-79.0 nM), and some of them also inhibited the epimastigotes growth of two T. cruzi strains in vivo.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein