Merck
  • Preparation of D-gulose from disaccharide lactitol using microbial and chemical methods.
Preparation of D-gulose from disaccharide lactitol using microbial and chemical methods.
Bioscience, biotechnology, and biochemistry (2013-02-09)
Kenji Morimoto, Tsuyoshi Shimonishi, Seiki Miyake, Goro Takata, Ken Izumori
ABSTRACT
When an M31 strain of Agrobacterium tumefaciens was grown in a mineral salt medium at 30 °C containing 1.0% lactitol as sole carbon source, a keto-sugar was efficiently accumulated in the supernatant. This oxidation from lactitol to the keto-sugar was caused by M31 cells grown with medium containing a disaccharide unit, including sucrose, lactitol, lactose, maltose, or maltitol, suggesting that the enzyme is inducible. M31 also demonstrated good growth characteristics in Tryptic Soy Broth (TSB) medium containing 1.0% sucrose, and cells grown under these conditions showed strong lactitol transformation activity. The keto-sugar product was reduced by chemical hydrogenation and the resulting product was hydrolyzed to D-gulose, D-galactose, and D-sorbitol by acid hydrolysis, revealing that the reduced products are lactitol and D-gulosyl-(β-1,4)-D-sorbitol. Taken together, these results indicate that M31 can convert lactitol to 3-ketolactitol and thus provide access to the rare sugar D-gulose.
Product Description
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Lactose (anhydrous), European Pharmacopoeia (EP) Reference Standard
Supelco
Lactose, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Maltitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Maltitol, ≥98% (HPLC)