Merck
Fluorescence emission properties of C60 nanoparticles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-02-12)
Fang Zhang, Yan Fang, Xiaoling Zhang
ABSTRACT
Fluorescence properties of C60 nanoparticles produced by sonication in pyridine have been systematically investigated. The fluorescence emission of C60 is dependent on the particle size. A red shift of emission maximum from 424 to 468 nm originates from the average size increase caused by strong interaction among C60 molecules in nanoparticles. In addition, it is also confirmed that the emergence of broad fluorescence emission band in the 550-570 nm region is ascribed to C60 nanoparticles aggregation.
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine hydrobromide, 98%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Supelco
Pyridine solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
Pyridine hydrochloride, purum, ≥98.0% (AT)