Merck
  • New hyaluronic acid based brush copolymers synthesized by atom transfer radical polymerization.
New hyaluronic acid based brush copolymers synthesized by atom transfer radical polymerization.
Carbohydrate polymers (2013-02-13)
Giovanna Pitarresi, Calogero Fiorica, Mariano Licciardi, Fabio Salvatore Palumbo, Gaetano Giammona
ABSTRACT
In this work, an efficient method for the synthesis of hyaluronic acid based brush copolymers using atom transfer radical polymerization (ATRP) has been reported. At first, two different hyaluronic acid (HA) based macroinitiators have been prepared and then they have been used for the polymerization via ATRP of hydrophilic or hydrophobic molecules carrying vinyl portions. In particular, by linking 2-bromo-2-methylpropionic acid (BMP) to the primary hydroxyl groups of tetrabutyl ammonium salt of HA (HA-TBA) or to amino groups of the ethylenediamino derivative of HA-TBA (HA-TBA-EDA), two macroinitiators (HA-TBA-BMP and HA-TBA-EDA-BMP) have been obtained. Then they have been used for the ATRP of poly(ethylene glycol) methacrylate (PEGMA), butyl methacrylate (BUTMA) or N-isopropylacrylamide (NIPAM) using a complex of Cu(I) and 2,2'-Bipyridyl (Bpy), as a catalyst. Both macroinitiators and final copolymers, named as HA-BMP-pPEGMA, HA-BMP-pBUTMA, HA-BMP-pNIPAM, HA-EDA-BMP-pPEGMA, HA-EDA-BMP-pBUTMA and HA-EDA-BMP-pNIPAM, have been characterized by spectroscopic analysis and size exclusion chromatography to confirm the success of the polymerization process.
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~98%
Sigma-Aldrich
Propionic acid, ≥99.5%