Merck
Interaction of vanadium(IV) with human serum apo-transferrin.
Journal of inorganic biochemistry (2013-02-16)
Sameena Mehtab, Gisela Gonçalves, Somnath Roy, Ana Isabel Tomaz, Teresa Santos-Silva, Marino F A Santos, Maria J Romão, Tamás Jakusch, Tamás Kiss, João Costa Pessoa
ABSTRACT
The interaction of V(IV)O-salts as well as of a few V(IV)O(carrier)n complexes with human serum transferrin (hTF) is studied focusing on the determination of the nature and stoichiometry of the binding of V(IV)O(2+) to hTF, as well as whether the conformation of hTF upon binding to V(IV)O(2+) or to its complexes is changed. Circular dichroism (CD) spectra measured for solutions containing V(IV)O(2+) and apo-hTF, and V(IV)O-maltol and apo-hTF, clearly indicate that hTF-V(IV)O-maltol ternary species form with a V(IV)O:maltol stoichiometry of 1:1. For V(IV)O salts and several V(IV)O(carrier)n complexes (carrier ligand=maltolato, dhp, picolinato and dipicolinato) (Hdhp=1,2-dimethyl-3-hydroxy-4-pyridinone) the maximum number of V(IV)O(2+) bound per mole of hTF is determined to be ~2 or lower in all cases. The binding of V(IV)O to apo-hTF most certainly involves several amino acid residues of the Fe-binding site, and as concluded by urea gel electrophoresis experiments, the formation of (V(IV)O)2hTF species may occur with the closing of the hTF conformation as is the case in (Fe(III))2hTF, which is an essential feature for the transferrin receptor recognition.
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
2-Picolinic acid, ReagentPlus®, 99%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
Maltol, natural, FG
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Supelco
2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate