Merck
  • Novel 3-O-carbamoyl erythromycin A derivatives (carbamolides) with activity against resistant staphylococcal and streptococcal isolates.
Novel 3-O-carbamoyl erythromycin A derivatives (carbamolides) with activity against resistant staphylococcal and streptococcal isolates.
Bioorganic & medicinal chemistry letters (2013-02-19)
Thomas V Magee, Seungil Han, Sandra P McCurdy, Thuy-Trinh Nguyen, Karl Granskog, Eric S Marr, Bruce A Maguire, Michael D Huband, Jinshan Michael Chen, Timothy A Subashi, Veerabahu Shanmugasundaram
ABSTRACT
A novel series of 3-O-carbamoyl erythromycin A derived analogs, labeled carbamolides, with activity versus resistant bacterial isolates of staphylococci (including macrolide and oxazolidinone resistant strains) and streptococci are reported. An (R)-2-aryl substituent on a pyrrolidine carbamate appeared to be critical for achieving potency against resistant strains. Crystal structures showed a distinct aromatic interaction between the (R)-2-aryl (3-pyridyl for 4d) substituent on the pyrrolidine and G2484 (G2505, Escherichia coli) of the Deinococcus radiodurans 50S ribosome (3.2Å resolution).
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyrrolidine, 99%
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N,N′-Bis(hydroxymethyl)urea, ≥95%