Merck
  • A nitrophenyl-based prodrug type for colorectal targeting of prednisolone, budesonide and celecoxib.
A nitrophenyl-based prodrug type for colorectal targeting of prednisolone, budesonide and celecoxib.
Bioorganic & medicinal chemistry letters (2013-02-19)
Juan F Marquez Ruiz, Kinga Kedziora, Maria Pigott, Brian Keogh, Henry Windle, Jason Gavin, Dermot P Kelleher, John F Gilmer
ABSTRACT
Celecoxib is a COX-2 inhibitor drug that can be used to reduce the risk of colorectal adenocarcinoma. Glucocorticoids are used in the treatment of inflammatory bowel disease. A limitation to the use of both drug types is that they undergo absorption from the intestinal tract with serious side effects. The prodrug systems introduced here involve forming a nitro-substituted acylsulfonamide group in the case of celecoxib and a nitro-substituted 21-ester for the glucocorticoids. Drug release is triggered by the nitro reductase action of the colonic microflora, liberating a cyclization competent species. The release of the active parent drugs was evaluated in vitro using Clostridium perfringens and epithelial transport through Caco-2 monolayer evaluation was carried out to estimate the absorption properties of the prodrugs compared to the parental drugs.
Product Description
Supelco
Budesonide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Budesonide, ≥99%
Budesonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%