Merck
  • Novel tryptophan metabolism by a potential gene cluster that is widely distributed among actinomycetes.
Novel tryptophan metabolism by a potential gene cluster that is widely distributed among actinomycetes.
The Journal of biological chemistry (2013-02-23)
Taro Ozaki, Makoto Nishiyama, Tomohisa Kuzuyama
ABSTRACT
The characterization of potential gene clusters is a promising strategy for the identification of novel natural products and the expansion of structural diversity. However, there are often difficulties in identifying potential metabolites because their biosynthetic genes are either silenced or expressed only at a low level. Here, we report the identification of a novel metabolite that is synthesized by a potential gene cluster containing an indole prenyltransferase gene (SCO7467) and a flavin-dependent monooxygenase (FMO) gene (SCO7468), which were mined from the genome of Streptomyces coelicolor A3(2). We introduced these two genes into the closely related Streptomyces lividans TK23 and analyzed the culture broths of the transformants. This process allowed us to identify a novel metabolite, 5-dimethylallylindole-3-acetonitrile (5-DMAIAN) that was overproduced in the transformant. Biochemical characterization of the recombinant SCO7467 and SCO7468 demonstrated the novel L-tryptophan metabolism leading to 5-DMAIAN. SCO7467 catalyzes the prenylation of L-tryptophan to form 5-dimethylallyl-L-tryptophan (5-DMAT). This enzyme is the first actinomycetes prenyltransferase known to catalyze the addition of a dimethylallyl group to the C-5 of tryptophan. SCO7468 then catalyzes the conversion of 5-DMAT into 5-dimethylallylindole-3-acetaldoxime (5-DMAIAOx). An aldoxime-forming reaction catalyzed by the FMO enzyme was also identified for the first time in this study. Finally, dehydration of 5-DMAIAOx presumably occurs to yield 5-DMAIAN. This study provides insight into the biosynthesis of prenylated indoles that have been purified from actinomycetes.
Sigma-Aldrich
2-Thiouracil, ≥99%
Sigma-Aldrich
3-Indoleacetonitrile, 98%