Merck
Atmospheric oxidation mechanism of phenol initiated by OH radical.
The journal of physical chemistry. A (2013-02-27)
Cui Xu, Liming Wang
ABSTRACT
The gas-phase oxidation mechanism of phenol initiated by OH radical was investigated using DFT and ab initio calculations. The initiation of the reaction is dominated by OH addition to ortho-position, forming P2, which subsequently combines with O2 at the ipso-position to form P2-1-OO adduct. A concerted HO2 elimination process from P2-1-OO was found to be much faster than the common ring closure to bicyclic intermediates. The HO2 elimination process from P2-1-OO forms 2-hydroxy-3,5-cyclohexadienone (HCH) as the main product and is also responsible for the experimental fact that the rate constants for reaction between P2 and O2 are about 2 orders of magnitude higher than those between other aromatic-OH adducts and O2. It was speculated that HCH would isomerize to catechol, which is thermodynamically more stable than HCH and was the experimentally observed main product, possibly through heterogeneous processes. Reaction of P2 with NO2 proceeded by addition to form P2-n-NO2 (n = 1, 3, 5), followed by HONO elimination from P2-1/3-NO2 to form catechol. The barriers for HONO elimination and catechol formation are below the separate reactants P2 and NO2, being consistent with the experimental observation of catechol in the absence of O2, while H2O elimination from P2-1/3-NO2 to form 2-nitrophenol (2NP) is hindered by high barriers. The most likely pathway for 2NP is the reaction of phenoxy radical and NO2.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
2-Nitrophenol, 98%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Pyrocatechol, ≥99%