Merck
  • Fortification of wheat flour and maize meal with different iron compounds: results of a series of baking trials.
Fortification of wheat flour and maize meal with different iron compounds: results of a series of baking trials.
Food and nutrition bulletin (2013-03-01)
Philip Randall, Quentin Johnson, Anna Verster
ABSTRACT
Wheat and maize flour fortification is a preventive food-based approach to improve the micronutrient status of populations. In 2009, the World Health Organization (WHO) released recommendations for such fortification, with guidelines on the addition levels for iron, folic acid, vitamin B12, vitamin A, and zinc at various levels of average daily consumption. Iron is the micronutrient of greatest concern to the food industry, as some believe there may be some adverse interaction(s) in some or all of the finished products produced from wheat flour and maize meal. To determine if there were any adverse interactions due to selection of iron compounds and, if differences were noted, to quantify those differences. Wheat flour and maize meal were sourced in Kenya, South Africa, and Tanzania, and the iron compound (sodium iron ethylenediaminetetraacetate [NaFeEDTA], ferrous fumarate, or ferrous sulfate) was varied and dosed at rates according to the WHO guidelines for consumption of 75 to 149 g/day of wheat flour and > 300 g/day of maize meal and tested again for 150 to 300 g/day for both. Bread, chapatti, ugali (thick porridge), and uji (thin porridge) were prepared locally and assessed on whether the products were acceptable under industry-approved criteria and whether industry could discern any differences, knowing that differences existed, by academic sensory analysis using a combination of trained and untrained panelists and in direct side-by-side comparison. Industry (the wheat and maize milling sector) scored the samples as well above the minimal standard, and under academic scrutiny no differences were reported. Side-by-side comparison by the milling industry did indicate some slight differences, mainly with respect to color, although these differences did not correlate with any particular iron compound. The levels of iron compounds used, in accordance with the WHO guidelines, do not lead to changes in the baking and cooking properties of the wheat flour and maize meal. Respondents trained to measure against a set benchmark and/or discern differences could not consistently replicate perceived difference observations.
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, suitable for plant cell culture, BioReagent, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)