Merck
Cytotoxic activities of some novel benzhydrylpiperazine derivatives.
Drug research (2013-03-01)
E E Gurdal, M Yarim, I Durmaz, R Cetin-Atalay
ABSTRACT
This study presents the synthesis of nineteen 1-(substitutedbenzoyl)-4-benzhydrylpiperazine and 1-[(substitutedphenyl)sulfonyl]-4-benzhydrylpiperazine derivatives. In vitro cytotoxic activities of the compounds were screened against hepatocellular (HUH-7), breast (MCF-7) and colorectal (HCT-116) cancer cell lines by sulphorhodamine B assay. Among the test compounds, benzamide derivatives had high cytotoxic activity whereas sulfonamide derivatives showed variable 50% growth inhibition (GI50).
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade