Merck
  • Gemcitabine-coumarin-biotin conjugates: a target specific theranostic anticancer prodrug.
Gemcitabine-coumarin-biotin conjugates: a target specific theranostic anticancer prodrug.
Journal of the American Chemical Society (2013-03-07)
Sukhendu Maiti, Nayoung Park, Ji Hye Han, Hyun Mi Jeon, Jae Hong Lee, Sankarprasad Bhuniya, Chulhun Kang, Jong Seung Kim
ABSTRACT
We present here, the design, synthesis, spectroscopic characterization, and in vitro biological assessment of a gemcitabine-coumarin-biotin conjugate (5). Probe 5 is a multifunctional molecule composed of a thiol-specific cleavable disulfide bond, a coumarin moiety as a fluorescent reporter, gemcitabine (GMC) as a model active drug, and biotin as a cancer-targeting unit. Upon addition of free thiols that are relatively abundant in tumor cells, disulfide bond cleavage occurs as well as active drug GMC release and concomitantly fluorescence intensity increases. Confocal microscopic experiments reveal that 5 is preferentially taken up by A549 cells rather than WI38 cells. Fluorescence-based colocalization studies using lysosome- and endoplasmic reticulum-selective dyes suggest that thiol-induced disulfide cleavage of 5 occur in the lysosome possibly via receptor-mediated endocytosis. The present drug delivery system is a new theranostic agent, wherein both a therapeutic effect and drug uptake can be readily monitored at the subcellular level by two photon fluorescence imaging.
Product Description
Biotin Alkyne, ≥95% (HPLC)
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Supelco
Coumarin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Coumarin, primary reference standard