Merck
Treating telangiectasias: my method.
Minerva cardioangiologica (2013-03-16)
F Ferrara, G Ferrara
ABSTRACT
The authors propose a new classification of telangiectasias: conditions involving demonstrated reflux are classified as type A telangiectasias; clustered, spider telangiectasias not related to reflux and with vein diameters of >0.2 mm are classified as type B, while isolated telangiectatic veins of ≤0.2 mm diameter are classed as type C. This histological and pathophysiological approach is the basis for the Authors' Multi-Therapy Treatment Protocol (MTT). The treatment regimen provides for initial treatment of type A telangiectasias with just conventional reflux sclerotherapy, followed three weeks later by treatment of type B telangiectasias with 0.25-0.5% polidocanol foam, associated with both external compression and tumescent vasoconstriction (START technique). This is then followed after a further three months by dermal stimulation with mesoglycan (LIDS technique) to reinforce the district underlying the type C telangiectasias. The MTT Protocol was used on 63 patients (125 limbs). A 12-month follow-up showed the treatment regimen to provide better aesthetic and functional results than classical sclerotherapy, with few adverse effects and greater patient satisfaction.
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O
Sigma-Aldrich
Thesit®
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Brij® L4, average Mn ~362
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography