Merck
  • Activated vibrational modes and Fermi resonance in tip-enhanced Raman spectroscopy.
Activated vibrational modes and Fermi resonance in tip-enhanced Raman spectroscopy.
Physical review. E, Statistical, nonlinear, and soft matter physics (2013-03-19)
Mengtao Sun, Yurui Fang, Zhenyu Zhang, Hongxing Xu
ABSTRACT
Using p-aminothiophenol (PATP) molecules on a gold substrate and high-vacuum tip-enhanced Raman spectroscopy (HV-TERS), we show that the vibrational spectra of these molecules are distinctly different from those in typical surface-enhanced Raman spectroscopy. Detailed first-principles calculations help to assign the Raman peaks in the TERS measurements as Raman-active and IR-active vibrational modes of dimercaptoazobenzene (DMAB), providing strong spectroscopic evidence for the dimerization of PATP molecules to DMAB under the TERS setup. The activation of the IR-active modes is due to enhanced electromagnetic field gradient effects within the gap region of the highly asymmetric tip-surface geometry. Fermi resonances are also observed in HV-TERS. These findings help to broaden the versatility of TERS as a promising technique for ultrasensitive molecular spectroscopy.
Sigma-Aldrich
Thiophenol, ≥99%
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Sigma-Aldrich
Thiophenol, 97%