Merck
  • Polyinosinic-polycytidylic acid is the most effective TLR adjuvant for SIV Gag protein-induced T cell responses in nonhuman primates.
Polyinosinic-polycytidylic acid is the most effective TLR adjuvant for SIV Gag protein-induced T cell responses in nonhuman primates.
Journal of immunology (Baltimore, Md. : 1950) (2013-03-20)
Haesun Park, Lauren Adamson, Tae Ha, Karl Mullen, Shoko I Hagen, Arys Nogueron, Andrew W Sylwester, Michael K Axthelm, Al Legasse, Michael Piatak, Jeffrey D Lifson, Juliana M McElrath, Louis J Picker, Robert A Seder
ABSTRACT
Prime-boost immunization with heterologous vaccines elicits potent cellular immunity. In this study, we assessed the influence of various TLR ligands on SIV Gag-specific T cell immunity and protection following prime-boost immunization. Rhesus macaques (RMs) were primed with SIV Gag protein emulsified in Montanide ISA51 with or without TLR3 (polyinosinic-polycytidylic acid [poly-IC]), TLR4 (monophosphoryl lipid A), TLR7/8 (3M-012), TLR9 (CpG), or TLR3 (poly-IC) combined with TLR7/8 ligands, then boosted with replication defective adenovirus 5 expressing SIV Gag (rAd5-Gag). After priming, RMs that received SIV Gag protein plus poly-IC developed significantly higher frequencies of SIV Gag-specific CD4(+) Th1 responses in blood and bronchoalveolar lavage (BAL) fluid lymphocytes compared with all other adjuvants, and low-level SIV Gag-specific CD8(+) T cell responses. After the rAd5-Gag boost, the magnitude and breadth of SIV Gag-specific CD8(+) T cell responses were significantly increased in RM primed with SIV Gag protein plus poly-IC, with or without the TLR7/8 ligand, or CpG. However, the anamnestic, SIV Gag-specific CD8(+) T cell response to SIVmac251 challenge was not significantly enhanced by SIV Gag protein priming with any of the adjuvants. In contrast, the anamnestic SIV Gag-specific CD4(+) T cell response in BAL was enhanced by SIV Gag protein priming with poly-IC or CpG, which correlated with partial control of early viral replication after SIVmac251 challenge. These results demonstrate that prime-boost vaccination with SIV Gag protein/poly-IC improves magnitude, breadth, and durability of CD4(+) T cell immune responses, which could have a role in the control of SIV viral replication.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination