Merck
  • No increase in bleeding identified in type 1 VWD subjects with D1472H sequence variation.
No increase in bleeding identified in type 1 VWD subjects with D1472H sequence variation.
Blood (2013-03-23)
Veronica H Flood, Kenneth D Friedman, Joan Cox Gill, Sandra L Haberichter, Pamela A Christopherson, Brian R Branchford, Raymond G Hoffmann, Thomas C Abshire, Amy L Dunn, Jorge A Di Paola, W Keith Hoots, Deborah L Brown, Cindy Leissinger, Jeanne M Lusher, Margaret V Ragni, Amy D Shapiro, Robert R Montgomery
ABSTRACT
The diagnosis of von Willebrand disease (VWD) is complicated by issues with current laboratory testing, particularly the ristocetin cofactor activity assay (VWF:RCo). We have recently reported a sequence variation in the von Willebrand factor (VWF) A1 domain, p.D1472H (D1472H), associated with a decrease in the VWF:RCo/VWF antigen (VWF:Ag) ratio but not associated with bleeding in healthy control subjects. This report expands the previous study to include subjects with symptoms leading to the diagnosis of type 1 VWD. Type 1 VWD subjects with D1472H had a significant decrease in the VWF:RCo/VWF:Ag ratio compared with those without D1472H, similar to the findings in the healthy control population. No increase in bleeding score was observed, however, for VWD subjects with D1472H compared with those without D1472H. These results suggest that the presence of the D1472H sequence variation is not associated with a significant increase in bleeding symptoms, even in type 1 VWD subjects.
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
SAFC
L-Histidine
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material