Merck
  • [Vasobral in the treatment of brain lesions: views of physicians and patients].
[Vasobral in the treatment of brain lesions: views of physicians and patients].
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova (2013-03-27)
P R Kamchatnov
ABSTRACT
A study was based on the survey of 419 neurologists and 1189 their patients with different forms of cerebrovascular diseases using a specially developed questionnaire. In most cases, vasobral was used as a monotherapy or in a complex treatment. Twenty-two percent of physicians reported that vasobral was the most effective compared to other drugs. The good tolerability of treatment (18%) and the broad spectrum of indications and clinical effects (17%) were reported as well. The large percentage (75%) of patients indicated the positive effect of vasobral on memory, reasoning, vertigo etc The maximal effect was identified in the treatment of mild cognitive impairment caused by chronic brain ischemia and vertebrobasilar insufficiency. Vasobral is recommended for a use in a complex therapy in patients with more severe brain lesions and cognitive deficit.
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard