Merck
  • Methionine salvage and S-adenosylmethionine: essential links between sulfur, ethylene and polyamine biosynthesis.
Methionine salvage and S-adenosylmethionine: essential links between sulfur, ethylene and polyamine biosynthesis.
The Biochemical journal (2013-03-29)
Margret Sauter, Barbara Moffatt, Maye Chin Saechao, Rüdiger Hell, Markus Wirtz
ABSTRACT
Both Met (methionine) and SAM (S-adenosylmethionine), the activated form of Met, participate in a number of essential metabolic pathways in plants. The subcellular compartmentalization of Met fluxes will be discussed in the present review with respect to regulation and communication with the sulfur assimilation pathway, the network of the aspartate-derived amino acids and the demand for production of SAM. SAM enters the ethylene, nicotianamine and polyamine biosynthetic pathways and provides the methyl group for the majority of methylation reactions required for plant growth and development. The multiple essential roles of SAM require regulation of its synthesis, recycling and distribution to sustain these different pathways. A particular focus of the present review will be on the function of recently identified genes of the Met salvage cycle or Yang cycle and the importance of the Met salvage cycle in the metabolism of MTA (5'-methylthioadenosine). MTA has the potential for product inhibition of ethylene, nicotianamine and polyamine biosynthesis which provides an additional link between these pathways. Interestingly, regulation of Met cycle genes was found to differ between plant species as shown for Arabidopsis thaliana and Oryza sativa.
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
SAFC
L-Methionine
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)