Merck
  • Aberrant activation of M phase proteins by cell proliferation-evoking carcinogens after 28-day administration in rats.
Aberrant activation of M phase proteins by cell proliferation-evoking carcinogens after 28-day administration in rats.
Toxicology letters (2013-03-29)
Atsunori Yafune, Eriko Taniai, Reiko Morita, Hitomi Hayashi, Kazuhiko Suzuki, Kunitoshi Mitsumori, Makoto Shibutani
ABSTRACT
We have previously reported that hepatocarcinogens increase liver cells expressing p21(Cip1), a G1 checkpoint protein and M phase proteins after 28-day treatment in rats. This study aimed to identify early prediction markers of carcinogens available in many target organs after 28-day treatment in rats. Immunohistochemical analysis was performed on Ki-67, p21(Cip1) and M phase proteins [nuclear Cdc2, phospho-Histone H3 (p-Histone H3), Aurora B and heterochromatin protein 1α (HP1α)] with carcinogens targeting different organs. Carcinogens targeting thyroid (sulfadimethoxine; SDM), urinary bladder (phenylethyl isothiocyanate), forestomach (butylated hydroxyanisole; BHA), glandular stomach (catechol; CC), and colon (2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine and chenodeoxycholic acid) were examined using a non-carcinogenic toxicant (caprolactam) and carcinogens targeting other organs as negative controls. All carcinogens increased Ki-67(+), nuclear Cdc2(+), p-Histone H3(+) or Aurora B(+) carcinogenic target cells, except for both colon carcinogens, which did not increase cell proliferation. On the other hand, p21(Cip1+) cells increased with SDM and CC. HP1α responded only to BHA. Results revealed carcinogens evoking cell proliferation concurrently induced cell cycle arrest at M phase or showing chromosomal instability reflecting aberration in cell cycle regulation, irrespective of target organs, after 28-day treatment. Therefore, M phase proteins may be early prediction markers of carcinogens evoking cell proliferation in many target organs.
Sigma-Aldrich
2-Phenylethyl isothiocyanate, FG
Supelco
Sulfadimethoxine, 98.0-102.0%
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Phenethyl isothiocyanate, 99%
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%