Merck
Unilateral lobar pulmonary edema.
The Journal of the Association of Physicians of India (2013-04-04)
Garvi J Pandya, Amartya Mukhopadhyay, Chua Ai Ping, Tow Keang Lim
ABSTRACT
Pulmonary edema is usually bilateral, but unilateral lobar pulmonary edema can also be encountered in clinical practice. We describe a case of unilateral lobar pulmonary edema in a patient without known cardiac history. It was first presentation of underlying cardiac disease in our patient and was difficult to differentiate from pneumonia. Unilateral pulmonary edema can mimic as pneumonia. Clinician should be aware of differential diagnosis of pulmonary edema, otherwise it can lead to unnecessary investigation and delay in starting definitive treatment.
Product Description
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide, Pharmaceutical Secondary Standard; Certified Reference Material
Furosemide, European Pharmacopoeia (EP) Reference Standard
Supelco
Furosemide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
Sigma-Aldrich
Furosemide