Merck
  • Urinary excretion of cyclohexanediol, a metabolite of the solvent cyclohexanone, by infants in a special care unit.
Urinary excretion of cyclohexanediol, a metabolite of the solvent cyclohexanone, by infants in a special care unit.
Clinical chemistry (1990-06-01)
G A Mills, V Walker
ABSTRACT
Using gas chromatography-mass spectrometry, we investigated the urinary excretion of organic acids of 278 newborn babies in a special care unit to obtain reference data and monitor metabolism. In 101 of 584 urine samples analyzed, we found isomers of cyclohexanediol. trans-1,2-Cyclohexanediol was always most abundant, with small amounts of 1,3- and 1,4-cyclohexanediol and, sometimes, traces of cis-1,2-cyclohexanediol. Glucuronide conjugates were not detected. The probable source was the solvent cyclohexanone, which was found as a contaminant of intravenous dextrose and the parenteral feeding solution, and was also leached into the infusion fluids from the administration set. We recovered 0.89 mg (range 0.74-0.98 mg, n = 5) of cyclohexanone from 150 mL of dextrose pumped through the infusion apparatus over 24 h, the normal rate for a 1-kg premature baby. Although this is well below toxic doses reported for mature animals, more data are needed for the newborn, particularly preterm infants who have a decreased capacity for glucuronide conjugation.
Sigma-Aldrich
trans-1,2-Cyclohexanediol, 98%
Sigma-Aldrich
cis-1,2-Cyclohexanediol, 99%
Sigma-Aldrich
1,4-Cyclohexanediol, 99%