Merck
Metabolic control of persister formation in Escherichia coli.
Molecular cell (2013-05-15)
Stephanie M Amato, Mehmet A Orman, Mark P Brynildsen
ABSTRACT
Bacterial persisters are phenotypic variants that form from the action of stress response pathways triggering toxin-mediated antibiotic tolerance. Although persisters form during normal growth from native stresses, the pathways responsible for this phenomenon remain elusive. Here we have discovered that carbon source transitions stimulate the formation of fluoroquinolone persisters in Escherichia coli. Further, through a combination of genetic, biochemical, and flow cytometric assays in conjunction with a mathematical model, we have reconstructed a molecular-level persister formation pathway from initial stress (glucose exhaustion) to the activation of a metabolic toxin-antitoxin (TA) module (the ppGpp biochemical network) resulting in inhibition of DNA gyrase activity, the primary target of fluoroquinolones. This pathway spans from initial stress to antibiotic target and demonstrates that TA behavior can be exhibited by a metabolite-enzyme interaction (ppGpp-SpoT), in contrast to classical TA systems that involve only protein and/or RNA.
Product Description
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Levofloxacin, 98.0-102.0% anhydrous basis (HPLC)
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Levofloxacin, analytical standard
Supelco
Ampicillin, analytical standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard