Merck
  • [Non-Hodgkin lymphoma of maxillofacial soft tissue: a report of two cases and review of the literature].
[Non-Hodgkin lymphoma of maxillofacial soft tissue: a report of two cases and review of the literature].
Cancer radiotherapie : journal de la Societe francaise de radiotherapie oncologique (2013-05-15)
S Chaabouni, H Ben Salah Lahiani, L Ghorbel, M Abdelmoula, M Elloumi, J Daoud
ABSTRACT
The extranodal non-Hodgkin lymphomas of maxillofacial soft tissues are rare. We report two cases of maxillofacial soft tissue non-Hodgkin lymphoma treated with chemotherapy followed by localized radiotherapy with complete remission after 3 and 6 months. We study the clinical, radiological and histopathological features as well as the treatment and the prognosis of extranodal non-Hodgkin lymphomas maxillofacial muscles.
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material