Merck
  • Aloe ferox seed: a potential source of oil for cosmetic and pharmaceutical use.
Aloe ferox seed: a potential source of oil for cosmetic and pharmaceutical use.
Natural product communications (2013-05-18)
Rachael Dangarembizi, Eliton Chivandi, Kennedy Erlwanger
ABSTRACT
Aloe ferox is an important medicinal plant in Southern Africa whose seeds could be useful as a source of oil. The fatty acid composition of A. ferox seed oil was determined using gas chromatography. The physicochemical properties of the oil were analysed using standard methods. The seeds yielded 19.4% of a light textured oil using the Blight and Dyer's method and 12.3% using the Soxhlet extraction method. The saponification value of the seed oil was 241.9 mg KOH/g and the peroxide value was 8.9 meq/kg. The acid value of the seed oil was 51.5 mg KOH/g (25.9% free fatty acids). The major fatty acids found in the seed oil were linoleic acid (71.8%), oleic acid (12.0%), palmitic acid (11.2%) and stearic acid (2.9%). The results obtained suggest that as A. ferox seed oil is high in linoleic acid, it could be potentially exploited in the cosmetic and pharmaceutical industries.
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Linoleic acid, analytical standard
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture