Merck
  • A posttranslational modification cascade involving p38, Tip60, and PRAK mediates oncogene-induced senescence.
A posttranslational modification cascade involving p38, Tip60, and PRAK mediates oncogene-induced senescence.
Molecular cell (2013-05-21)
Hui Zheng, Alim Seit-Nebi, Xuemei Han, Aaron Aslanian, John Tat, Rong Liao, John R Yates, Peiqing Sun
ABSTRACT
Oncogene-induced senescence is an important tumor-suppressing defense mechanism. However, relatively little is known about the signaling pathway mediating the senescence response. Here, we demonstrate that a multifunctional acetyltransferase, Tip60, plays an essential role in oncogenic ras-induced senescence. Further investigation reveals a cascade of posttranslational modifications involving p38, Tip60, and PRAK, three proteins that are essential for ras-induced senescence. Upon activation by ras, p38 induces the acetyltransferase activity of Tip60 through phosphorylation of Thr158; activated Tip60 in turn directly interacts with and induces the protein kinase activity of PRAK through acetylation of K364 in a manner that depends on phosphorylation of both Tip60 and PRAK by p38. These posttranslational modifications are critical for the prosenescent function of Tip60 and PRAK, respectively. These results have defined a signaling pathway that mediates oncogene-induced senescence, and identified posttranslational modifications that regulate the enzymatic activity and biological functions of Tip60 and PRAK.
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Threonine
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein