Merck
  • Hypertonic sodium bicarbonate versus intravenous lipid emulsion in a rabbit model of intravenous flecainide toxicity: no difference, no sink.
Hypertonic sodium bicarbonate versus intravenous lipid emulsion in a rabbit model of intravenous flecainide toxicity: no difference, no sink.
Clinical toxicology (Philadelphia, Pa.) (2013-05-25)
G Cave, M Harvey, P Quinn, D Heys
ABSTRACT
The use of intravenous lipid emulsion (ILE) as an antidote in non-local, anaesthetic drug toxicity has generated considerable interest. Flecainide is a lipophilic anti-arrhythmic with a significant cardiotoxic profile, with blockade of sodium and potassium channels causing arrhythmias and shock in severe toxicity. ILE has been proposed as a treatment option in severe flecainide toxicity refractory to other modalities. We compared the effects of ILE and hypertonic sodium bicarbonate in a rabbit model of flecainide toxicity. Twenty sedated and ventilated New Zealand White Rabbits received flecainide infusion titrated to a mean arterial pressure (MAP) of 60% baseline, which was defined as toxicity. The rabbits then received either sodium bicarbonate or ILE, and the flecainide infusion was reduced in an attempt to model ongoing enteric absorption. MAP and heart rate were recorded every minute for 15 min and plasma flecainide concentration was measured at toxicity and 15 min. ECG QRS duration was recorded at baseline, toxicity and at 5, 10 and 15 min post-toxicity. No difference was observed in heart rate (p = 0.2804), MAP (p = 0.1802) or QRS duration (p = 0.7471) between groups. The immediate rate of rise in MAP was greatest in the bicarbonate group in the 5 min immediately post-toxicity. In this study, no differences were observed between an active control of hypertonic sodium bicarbonate and ILE for the primary endpoint of MAP at 15 min nor for QRS duration at any timepoint. There was a transient rapid increase in blood pressure seen in the sodium bicarbonate group that was not sustained. No increase was seen in blood concentration of flecainide in the ILE group, suggesting no 'lipid sink' for flecainide in this model. More research is warranted to define any role for ILE in flecainide toxicity.
Product Description
Supelco
(±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Flecainide acetate salt
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C