Merck
TP53 and outcome in DLBCL: not only the coding region.
Blood (2013-06-01)
Fabrice Jardin, Bertrand Coiffier
ABSTRACT
In this issue of Blood, Li et al report mutations in the 3′ untranslated region (3′UTR) of TP53 that modify the expression of p53 and thus its effect on response to therapy in diffuse large B-cell lymphoma (DLBCL) patients.
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%