Merck
  • Electronic monitoring of oral therapies in ethnically diverse and economically disadvantaged patients with rheumatoid arthritis: consequences of low adherence.
Electronic monitoring of oral therapies in ethnically diverse and economically disadvantaged patients with rheumatoid arthritis: consequences of low adherence.
Arthritis and rheumatism (2013-06-04)
Christian A Waimann, Maria F Marengo, Sofia de Achaval, Vanessa L Cox, Araceli Garcia-Gonzalez, John D Reveille, Marsha N Richardson, Maria E Suarez-Almazor
ABSTRACT
To quantify adherence to oral therapies in ethnically diverse and economically disadvantaged patients with rheumatoid arthritis (RA), using electronic medication monitoring, and to evaluate the clinical consequences of low adherence. A total of 107 patients with RA enrolled in a 2-year prospective cohort study agreed to have their oral RA drug therapy intake electronically monitored using the Medication Event Monitoring System. Adherence to disease-modifying antirheumatic drugs (DMARDs) and prednisone was determined as the percentage of days (or weeks for methotrexate) on which the patient took the correct dose as prescribed by the physician. Patient outcomes were assessed, including function measured by the modified Health Assessment Questionnaire, disease activity measured by the Disease Activity Score in 28 joints (DAS28), health-related quality of life, and radiographic damage measured using the modified Sharp/van der Heijde scoring method. Adherence to the treatment regimen as determined by the percentage of correct doses was 64% for DMARDs and 70% for prednisone. Patients who had better mental health were statistically more likely to be adherent. Only 23 of the patients (21%) had an average adherence to DMARDs ≥80%. These patients showed significantly better mean DAS28 values across 2 years of followup than those who were less adherent (3.28 versus 4.09; P = 0.02). Radiographic scores were also worse in nonadherent patients at baseline and at 12 months. Only one-fifth of RA patients had an overall adherence to DMARDs of at least 80%. Less than two-thirds of the prescribed DMARD doses were correctly taken. Adherent patients had lower disease activity across the 2 years of followup.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate, European Pharmacopoeia (EP) Reference Standard