Merck
  • High dietary levels of biotin and zinc to improve health of foot pads in broilers exposed experimentally to litter with critical moisture content.
High dietary levels of biotin and zinc to improve health of foot pads in broilers exposed experimentally to litter with critical moisture content.
Poultry science (2013-06-19)
A Abd El-Wahab, D Radko, J Kamphues
ABSTRACT
Foot pad dermatitis (FPD) is a widespread problem in poultry production and constitutes a welfare issue. The objective of this study was to test potentially prophylactic effects of higher biotin and Zn levels in the diet of broilers exposed to critical litter moisture content (35% water) on the development of FPD. Two trials were performed in each 4 groups of 1-wk-old male broilers (Ross 708) during 33 d. The pens of all groups (25 birds in each) were littered with wood shavings of critical moisture content. Two groups were fed high levels of Zn as zinc-oxide (150 mg/kg of diet), with normal levels of biotin (300 µg/kg of diet) or high biotin (2,000 µg/kg of diet). The other 2 groups were fed Zn as zinc-methionine (150 mg/kg of diet), with normal levels of biotin (300 µg/kg of diet) or high biotin (2,000 µg/kg of diet). External assessment of foot pads and measurements the moisture contents of excreta and litter were performed weekly. The signs of foot pad lesions were recorded on a 7-point scale (0 = normal skin; 7 = more than half of the foot pad is necrotic). High biotin supplementation resulted in a reduction of 30 and 18% of cases of foot pad lesions in trials 1 and 2, respectively. The combination of Zn-methionine and high biotin supplementation led to a decreased severity of FPD in a range of about 50 and 30% in trials 1 and 2, respectively. In broilers fed the diet containing zinc-oxide and normal biotin levels about 28 and 24% of the birds had the scores of 6 and 7 (= high foot pad alterations), whereas in birds fed Zn-methionine and high biotin no high alterations (score = 7) in the foot pad (0%) occurred in either trial. The presented results suggest that it is advisable to combine the maximum levels of Zn (especially of Zn-methionine) and high levels of biotin when clinically relevant alterations in the foot pad occur.
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
SAFC
L-Methionine
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland