Merck
  • Kinetics of reversible reductive carbonylation of heme in human cystathionine β-synthase.
Kinetics of reversible reductive carbonylation of heme in human cystathionine β-synthase.
Biochemistry (2013-06-25)
Sebastián Carballal, Ernesto Cuevasanta, Inés Marmisolle, Omer Kabil, Carmen Gherasim, David P Ballou, Ruma Banerjee, Beatriz Alvarez
ABSTRACT
Cystathionine β-synthase (CBS) catalyzes the condensation of homocysteine with serine or cysteine to form cystathionine and water or hydrogen sulfide (H2S), respectively. In addition to pyridoxal phosphate, human CBS has a heme cofactor with cysteine and histidine as ligands. While Fe(III)-CBS is inert to exogenous ligands, Fe(II)-CBS can be reversibly inhibited by carbon monoxide (CO) and reoxidized by O2 to yield superoxide radical. In this study, we have examined the kinetics of Fe(II)CO-CBS formation and reoxidation. Reduction of Fe(III)-CBS by dithionite showed a square root dependence on concentration, indicating that the reductant species was the sulfur dioxide radical anion (SO2(•-)) that exists in rapid equilibrium with S2O4(2-). Formation of Fe(II)CO-CBS from Fe(II)-CBS and 1 mM CO occurred with a rate constant of (3.1 ± 0.4) × 10(-3) s(-1) (pH 7.4, 25 °C). The reaction of Fe(III)-CBS with the reduced form of the flavoprotein methionine synthase reductase in the presence of CO and NADPH resulted in its reduction and carbonylation to form Fe(II)CO-CBS. Fe(II)-CBS was formed as an intermediate with a rate constant of (9.3 ± 2.5) × 10(2) M(-1) s(-1). Reoxidation of Fe(II)CO-CBS by O2 was multiphasic. The major phase showed a hyperbolic dependence on O2 concentration. Although H2S is a product of the CBS reaction and a potential heme ligand, we did not find evidence of an effect of exogenous H2S on activity or heme binding. Reversible reduction of CBS by a physiologically relevant oxidoreductase is consistent with a regulatory role for the heme and could constitute a mechanism for cross talk among the CO, H2S, and superoxide signaling pathways.
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Histidine
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Wacker Chemie AG, 98.5-101.0%
Sigma-Aldrich
Sulfur dioxide, ≥99.9%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Supelco
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard