Merck
  • Separation of delta6- and delta9,11-estradiol: analytical method development, validation and practical application.
Separation of delta6- and delta9,11-estradiol: analytical method development, validation and practical application.
Die Pharmazie (2013-06-28)
K Schulz, U Oberdieck, W Weitschies
ABSTRACT
For estradiol (E2) the separation of the degradation products delta6- and delta9,11-E2 is especially challenging due to their structural similarity. There is no method described in the literature yet which adequately addresses this problem. The present study describes a HPLC method for the separation and quantitation of E2 and its degradation products 6alpha-hydroxy-E2, 6beta-hydroxy-E2, 6-keto-E2, delta9,11-E2, beta-equilenol and delta6-E2. The method employs a Kinetex PFP analytical column, using methanol and deionized water as mobile phases. Different UV- and fluorescence detection modes were used for maximal sensitivity and specificity. The applicability and capability of the method was demonstrated for Vagifem tablets. Finally, the method was validated with respect to selectivity, sensitivity, linearity, precision and accuracy.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.