Merck
  • [(-)-Epigallocatechin gallate regulates expression of apoptotic genes and protects cultured human lens epithelial cells under hyperglycemia].
[(-)-Epigallocatechin gallate regulates expression of apoptotic genes and protects cultured human lens epithelial cells under hyperglycemia].
Molekuliarnaia biologiia (2013-07-03)
Panpan Ye, Kana Lin, Zhaochun Li, Jian Liu, Ke Yao, Wen Xu
ABSTRACT
(-)-Epigallocatechin gallate (EGCG), the most abundant component in green tea, has a potent anti-apoptotic activity. The purpose of this study was to investigate the protective effects of EGCG and their molecular mechanisms on high glucose-induced apoptosis of human lens epithelial cells (HLEB-3). HLEB-3 cells were exposed to various concentrations of glucose and EGCG. Cell death was assessed by MTT assay and flow cytometry using annexin V and propidium iodide. The expression of the Bcl-2 family, c-fos, c-myc and p53 was measured by real-time PCR. EGCG decreased the Bcl-2/Bax expression stimulated by a high glucose. Moreover, EGCG suppressed the high glucose-induced expression of c-fos, c-myc and p53. These findings suggest that EGCG protects HLEB-3 cells from high glucose-induced apoptosis by regulating the gene expression of the Bcl-2 family, c-fos, c-myc and p53. Thus, EGCG may have a potential protective effect against diabetic cataract formation.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(+)-Catechin, analytical standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Epigallocatechin gallate, primary reference standard
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Epicatechin, primary reference standard