Merck
  • A thiophene-containing compound as a matrix for matrix-assisted laser desorption/ ionization mass spectrometry and the electrical conductivity of matrix crystals.
A thiophene-containing compound as a matrix for matrix-assisted laser desorption/ ionization mass spectrometry and the electrical conductivity of matrix crystals.
European journal of mass spectrometry (Chichester, England) (2013-07-12)
Akikazu Yasuda, Takayuki Ishimaru, Shogo Nishihara, Masamichi Sakai, Hideya Kawasaki, Ryuichi Arakawa, Yasushi Shigeri
ABSTRACT
The electrical conductivity of the matrix crystal might be a new factor to enhance matrix-assisted laser desorption/ionization mass spectrometry (MALDI-MS) sensitivity. In MALDI-MS, several compounds are used as a standard matrix. Utilization of such compounds is based on an a posteriori approach, but there is no theoretical guidance for selecting a matrix. In an attempt to further understand performance in MALDI-MS, we utilized peptide detection for random screening of a chemical library (12,383 compounds) for compounds with matrix functions in MALDI-MS. A lot of thiophene compounds were found to be a matrix, in which 2-[5-(2,4-dichlorobenzoyl)-2-thienyl] acetic acid (DCBTA) provided an important clue to measure the electrical conductivity of the matrix crystal, because the structure of DCBTA is analogous to conductive polymers and organic solar cells. Most of the crystals of standard matrices, such as alpha-cyano-4-hydroxycinnamic acid (CHCA), 3,5-dimethoxy-4-hydroxycinnamic acid [sinapinic acid, (SA)], and DCBTA showed electrical conductivity, whereas the conductivity of crystal was not observed in 2,5-dihydroxybenzoic acid (2,5-DHB). On the other hand, super-DHB using 2-hydroxy-5-methoxybenzoic acid [5-methoxysalicylic acid, (MSA)] as an additive to 2,5-DHB, improved the electrical conductivity of the crystal, that followed the enhancement of peak intensity in MS spectrum. These observations might indicate that the electrical conductivity of matrix crystals is a key consideration in obtaining efficient MALDI performance.
Product Description
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
2-Hydroxy-5-methoxybenzoic acid, 98%
Supelco
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for matrix substance for MALDI-MS, Ultra pure
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS