Merck
  • Solvothermal fabrication of rubidium tungsten bronze for the absorption of near infrared light.
Solvothermal fabrication of rubidium tungsten bronze for the absorption of near infrared light.
Journal of nanoscience and nanotechnology (2013-07-19)
Chongshen Guo, Shu Yin, Qiang Dong, Takeshi Kimura, Miyuki Tanaka, Le Thi Hang, Xiaoyong Wu, Tsugio Sato
ABSTRACT
For fabricating the crystal of Rb(x)WO3, the solvothermal process was performed using a mixed medium of water and ethanol as solvent. It has been found that the morphological evolutions of samples were greatly changed as disordered nanoparticles, bundle of nanorods and microfibers with increasing content of water in the starting solution. Among them, sample synthesized with assistance of 3 ml water consisted of the nanorods with a diameter in the range of 80-150 nm, showing suitable size and morphology as near-infrared shielding material. On the merits of this nanostructure, the thin film of the Rb(x)WO3 nanorods realized excellent shielding of near-infrared light, meanwhile, it selectively transmitted the major part of visible light.
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tin, ≥99%, powder
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®