Merck
  • Numerical simulation of solid liquid interface behavior during continuous strip casting process.
Numerical simulation of solid liquid interface behavior during continuous strip casting process.
Journal of nanoscience and nanotechnology (2013-07-19)
Changbum Lee, Wooyoung Yoon, Seungwon Shin, Jaewoo Lee, Bo-Yun Jang, Joonsoo Kim, Youngsoo Ahn, Jinseok Lee
ABSTRACT
A new metal-strip-casting process called continuous strip-casting (CSC) has been developed for making thin metal strips. A numerical simulation model to help understand solid-liquid interface behavior during CSC has been developed and used to identify the solidification morphologies of the strips and to determine the optimum processing conditions. In this study, we used a modified level contour reconstruction method (LCRM) and the sharp interface method to modify interface tracking, and performed a simulation analysis of the CSC process. The effects of process parameters such as heat-transfer coefficient and extrusion velocity on the behavior of the solid-liquid interface were estimated and used to improve the apparatus. A Sn (Tin) plate of dimensions 200 x 50 x 1 mm3 was successfully produced by CSC for a heat-transfer coefficient of 104 W/m2 K and an extrusion velocity of 0.2 m/s.
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis