Merck
  • Vasopressin, steroids, and epinephrine and neurologically favorable survival after in-hospital cardiac arrest: a randomized clinical trial.
Vasopressin, steroids, and epinephrine and neurologically favorable survival after in-hospital cardiac arrest: a randomized clinical trial.
JAMA (2013-07-19)
Spyros D Mentzelopoulos, Sotirios Malachias, Christos Chamos, Demetrios Konstantopoulos, Theodora Ntaidou, Androula Papastylianou, Iosifinia Kolliantzaki, Maria Theodoridi, Helen Ischaki, Dimosthemis Makris, Epaminondas Zakynthinos, Elias Zintzaras, Sotirios Sourlas, Stavros Aloizos, Spyros G Zakynthinos
ABSTRACT
Among patients with cardiac arrest, preliminary data have shown improved return of spontaneous circulation and survival to hospital discharge with the vasopressin-steroids-epinephrine (VSE) combination. To determine whether combined vasopressin-epinephrine during cardiopulmonary resuscitation (CPR) and corticosteroid supplementation during and after CPR improve survival to hospital discharge with a Cerebral Performance Category (CPC) score of 1 or 2 in vasopressor-requiring, in-hospital cardiac arrest. Randomized, double-blind, placebo-controlled, parallel-group trial performed from September 1, 2008, to October 1, 2010, in 3 Greek tertiary care centers (2400 beds) with 268 consecutive patients with cardiac arrest requiring epinephrine according to resuscitation guidelines (from 364 patients assessed for eligibility). Patients received either vasopressin (20 IU/CPR cycle) plus epinephrine (1 mg/CPR cycle; cycle duration approximately 3 minutes) (VSE group, n = 130) or saline placebo plus epinephrine (1 mg/CPR cycle; cycle duration approximately 3 minutes) (control group, n = 138) for the first 5 CPR cycles after randomization, followed by additional epinephrine if needed. During the first CPR cycle after randomization, patients in the VSE group received methylprednisolone (40 mg) and patients in the control group received saline placebo. Shock after resuscitation was treated with stress-dose hydrocortisone (300 mg daily for 7 days maximum and gradual taper) (VSE group, n = 76) or saline placebo (control group, n = 73). Return of spontaneous circulation (ROSC) for 20 minutes or longer and survival to hospital discharge with a CPC score of 1 or 2. Follow-up was completed in all resuscitated patients. Patients in the VSE group vs patients in the control group had higher probability for ROSC of 20 minutes or longer (109/130 [83.9%] vs 91/138 [65.9%]; odds ratio [OR], 2.98; 95% CI, 1.39-6.40; P = .005) and survival to hospital discharge with CPC score of 1 or 2 (18/130 [13.9%] vs 7/138 [5.1%]; OR, 3.28; 95% CI, 1.17-9.20; P = .02). Patients in the VSE group with postresuscitation shock vs corresponding patients in the control group had higher probability for survival to hospital discharge with CPC scores of 1 or 2 (16/76 [21.1%] vs 6/73 [8.2%]; OR, 3.74; 95% CI, 1.20-11.62; P = .02), improved hemodynamics and central venous oxygen saturation, and less organ dysfunction. Adverse event rates were similar in the 2 groups. Among patients with cardiac arrest requiring vasopressors, combined vasopressin-epinephrine and methylprednisolone during CPR and stress-dose hydrocortisone in postresuscitation shock, compared with epinephrine/saline placebo, resulted in improved survival to hospital discharge with favorable neurological status. clinicaltrials.gov Identifier: NCT00729794.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Adrenaline, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Methylprednisolone, European Pharmacopoeia (EP) Reference Standard