Merck
  • Swift adsorptive removal of Congo red from aqueous solution by K1.33Mn8O16 nanowires.
Swift adsorptive removal of Congo red from aqueous solution by K1.33Mn8O16 nanowires.
Journal of nanoscience and nanotechnology (2013-07-26)
Junshu Wu, Hongyi Li, Jinshu Wang, Zhifei Li
ABSTRACT
A swift and efficient approach to converting organic dye effluents into fresh water could be of substantial benefit. In this study, we presented facile hydrothermal synthesis of K1.33Mn8O16 nanowires in ammonium fluoride (NH4F) aqueous solution. The crystallization process of K1.33Mn8O16 nanowires was investigated. The as-obtained K1.33Mn8O16 nanowires were used for swift adsorptive removal of Congo red from aqueous solution without adjusting pH value at room temperature. Adsorption kinetic experimental data are well described by pseudo-second-order rate kinetic model, and the adsorption isotherm fits Langmuir isotherm model. The present investigation provides an efficient approach to designing and fabricating manganese-based nanomaterials for environmental remediation.
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Congo Red, analytical standard
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Congo Red, Dye content ≥35 %
Sigma-Aldrich
Congo Red, certified by the BSC, BioXtra
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%