Merck
  • Interaction between cochleata and stipule-reduced mutations results in exstipulate hypertrophied leaves in Pisum sativum L.
Interaction between cochleata and stipule-reduced mutations results in exstipulate hypertrophied leaves in Pisum sativum L.
Indian journal of experimental biology (2013-08-01)
Arvind Kumar, Vishakha Sharma, Sushil Kumar
ABSTRACT
In the wild type P. sativum, each of the adult plant stem nodes, bears a pair of sessile foliaceous stipules and a petiolated unipinnately compound leaf of 4 to 6 leaflets and 7-9 tendrils. The stipule-reduced (st) and cochleata (coch) single null mutants and coch st double null mutant differ fom the wild type in respectively having sessile stipules of much reduced size, petiolated simple and/or compound leaf-like stipules and no stipules. It is also known that coch leaves are somewhat bigger than st and wild type leaves. Here, pleiotropic phenotype of coch st double mutant was investigated. The morphologies of stipules and leaf were quantified in the field grown plants and microcultured shoots, latter in the presence and absence of gibberellic acid and N-1-naphthylphthalamic acid. The observations showed that as compared to the corresponding plants or shoots of COCH ST (WT) genotype, (a) coch st plants bore leaves in which all the organs were hypertrophied; (b) full complement of leaflets and 3-5 tendrils were formed on leaf; (c) the microcultured coch st shoots were taller despite lower number of nodes, and (d) they also produced leaves in which all the organs were bigger and the ratio of leaflets/tendrils was higher. It was concluded that in coch st double mutant (a) ST function is essential for stipule primordium differentiation, in the absence of COCH function and (b) absence of negative feedback loops between simple stipules and compound leaf for metabolite utilization allows hypertrophied growth in leaves.
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
Supelco
Gibberellic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)