Merck
  • Sorption and transport studies of cetyl trimethylammonium bromide (CTAB) and Triton X-100 in clayey soil.
Sorption and transport studies of cetyl trimethylammonium bromide (CTAB) and Triton X-100 in clayey soil.
Journal of environmental sciences (China) (2013-08-08)
Sivaram Harendra, Cumaraswamy Vipulanandan
ABSTRACT
Surfactants are soil washing agents and facilitators for subsurface remediation of hydrocarbon spills. It is important to understand the sorption and transport behavior of surfactants for enhanced soil remediation. The adsorption and desorption isotherms of cetyl trimethylammonium bromide (CTAB) and Triton X-100 with sand and kaolinite have been quantified. Kaolinite clay had the highest sorption capacity compared to blasting sand. Transport parameters such as diffusion coefficient (D) and retardation factor (R) of the above mentioned surfactant solutions were determined in clayey soils (82.5% sand and 17.5% kaolinite mixture) with near zero and 0.1 g/L ionic strength. NaCl was used as the electrolyte solution. Convection-Diffusion equation was used to model the breakthrough curves of the surfactants. Bromide ion was chosen as the tracer material in order to characterize the column. CTAB and Triton X-100 were used to flush the perchloroethylene (PCE) contaminated soil. The effectiveness of CTAB and Triton X-100 in flushing the PCE from the contaminated soil was quantified.
Product Description
Octoxinol 10, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triton X-102
Supelco
Tetrachloroethylene, analytical standard
Sigma-Aldrich
Triton X-305 solution, 70% in H2O
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Triton X-100 solution, BioUltra, Molecular Biology, ~10% in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Tetrachloroethylene, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrachloroethylene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Halloysite nanoclay
Supelco
Density Standard 1623 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Kaolin
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-45
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)